Diisoamylether (mix af isomere) ≥98.0% (by GC) stabilised

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel


Stabilization: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)

I0086-25ML I0086-500ML
TCIAI0086-25MLEA 132 DKK
TCIAI0086-25ML TCIAI0086-500ML
Diisoamylether (mix af isomere) ≥98.0% (by GC) stabilised
Diisoamylether (mix af isomere)
Formel: C₁₀H₂₂O
Molekylvægt: 158.28 g/mol
Kogepunkt: 173°C
Flammepunkt: 48°C
MDL Number: MFCD00008947
CAS-nummer: 544-01-4
EINECS: 208-857-4
UN nummer: 3271
ADR: 3 III
Merck Index: 14,05118
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=62fb20b3780f2691b55992319ac34d8b7111ff8e&vid=62fb20b3780f2691b55992319ac34d8b7111ff8e&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR