Cyclopentadienylthallium ≥95.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Thallium(I) cyclopentadienide

C0954-5G C0954-1G
TCIAC0954-5GEA 2430 DKK
TCIAC0954-5G TCIAC0954-1G
Cyclopentadienylthallium ≥95.0% (by titrimetric analysis)
Cyclopentadienylthallium
Formel: C₅H₅Tl
Molekylvægt: 269.48 g/mol
Smeltepunkt: 75
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00134141
CAS-nummer: 34822-90-7
EINECS: 252-229-2
UN nummer: 1707
ADR: 6.1 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=90a7ff8986b3e836ee7b3fb77a0b180461c194dd&vid=90a7ff8986b3e836ee7b3fb77a0b180461c194dd&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (Chelometric Titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR