Bis(triphenylphosphin)palladium(II)chlorid ≥98.0% (by titrimetric analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
Bis(triphenylphosphine)palladium(II) dichloride
B1667-1G
B1667-25G
B1667-5G
TCIAB1667-1GEA
427
DKK
TCIAB1667-1G
TCIAB1667-25G
TCIAB1667-5G
Bis(triphenylphosphin)palladium(II)chlorid ≥98.0% (by titrimetric analysis)
Bis(triphenylphosphin)palladium(II)chlorid
Formel:
C₃₆H₃₀Cl₂P₂Pd Molekylvægt: 701.91 g/mol Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD00009593 CAS-nummer: 13965-03-2 EINECS: 237-744-2 |
|
Specifikationer Testresultater
Appearance | Light yellow to Brown powder to crystal |
Purity (Chelometric Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...