5-Formyl-2-thiophenboronsyre (contains varying amounts of Anhydride)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 5-Formylthiophen-2-boronic acid, 5-Formylthiophene-2-boronic acid

F0549-1G F0549-5G
TCIAF0549-1GEA 148 DKK
TCIAF0549-1G TCIAF0549-5G
5-Formyl-2-thiophenboronsyre (contains varying amounts of Anhydride)
5-Formyl-2-thiophenboronsyre
Formel: C₅H₅BO₃S
Molekylvægt: 155.97 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD02093666
CAS-nummer: 4347-33-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=953259747aa6e2eb98fb0a5c55cc7df1fd3a509b&vid=953259747aa6e2eb98fb0a5c55cc7df1fd3a509b&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 113.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR