Tetrabutylammoniumfluorid ∼1 mol/L in THF
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
T1338-100ML
T1338-25ML
T1338-500ML
TCIAT1338-100MLEA
501
DKK
TCIAT1338-100ML
TCIAT1338-25ML
TCIAT1338-500ML
Tetrabutylammoniumfluorid ∼1 mol/L in THF
Tetrabutylammonium fluoride
Formel:
C₁₆H₃₆FN Molekylvægt: 261.47 g/mol Massefylde: 0.903 Flammepunkt: –17°C Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD00011747 CAS-nummer: 429-41-4 EINECS: 207-057-2 UN nummer: 2924 ADR: 3 II Merck Index: 14,09187 |
|
Specifikationer Testresultater
Appearance | Colorless to Red to Green clear liquid |
Concentration (Nonaqueous Titration) | 1.0 to 1.1 mol/L |
Water | max. 10.0 % |
TBHF-HF (neutralization titration) | max. 5.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...