2-Iodtoluen ≥98.0% (by GC) stabilised
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
1-Iodo-2-methylbenzene
Stabilization:
Stabilised with copper chip
I0217-25G
I0217-250G
TCIAI0217-25GEA
296
DKK
TCIAI0217-25G
TCIAI0217-250G
2-Iodtoluen ≥98.0% (by GC) stabilised
2-Iodtoluen
Formel:
C₇H₇I Molekylvægt: 218.04 g/mol Kogepunkt: 211°C Flammepunkt: 90 Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00001042 CAS-nummer: 615-37-2 EINECS: 210-422-9 |
|
Specifikationer Testresultater
Appearance | Colorless to Red to Green clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...