Methylboronsyre (contains varying amounts of Anhydride)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

M1553-1G M1553-5G
TCIAM1553-1GEA 140 DKK
TCIAM1553-1G TCIAM1553-5G
Methylboronsyre (contains varying amounts of Anhydride)
Methylboronsyre
Formel: CH₅BO₂
MDL Number: MFCD00002105
CAS-nummer: 13061-96-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1a48bfc4b28dcae3a6b3a6c45202c7846016a744&vid=1a48bfc4b28dcae3a6b3a6c45202c7846016a744&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow powder to crystal
Purity (Neutralization titration) 97.0 to 143.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR