5-Amino-2-hydroxybenzoesyre ≥98.0% (by titrimetric analysis) for biochemical research

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Mesalamine, Mesalazine, 5-Amino-2-hydroxybenzoic acid, m-Aminosalicylic acid

A2291-25G A2291-5G
TCIAA2291-25GEA 534 DKK
TCIAA2291-25G TCIAA2291-5G
5-Amino-2-hydroxybenzoesyre ≥98.0% (by titrimetric analysis) for biochemical research
5-Amino-2-hydroxybenzoesyre
Formel: C₇H₇NO₃
Molekylvægt: 153.14 g/mol
Kogepunkt: 347
Smeltepunkt: 265°C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00007877
CAS-nummer: 89-57-6
EINECS: 201-919-1
Merck Index: 14,05904
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=958279d533fa515523ac6cc3fa91ac73180edc8a&vid=ad7891cbc4b1a3467fa863ad3c0bb832592042ff&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light red to Green powder to crystal
Purity (Neutralization titration) min. 98.0 %
Hydrogendioxide detection test (Peroxidase method) to pass test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR