1-Brom-2-iodbenzen ≥98.0% (by GC) stabilised
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
2-Bromo-1-iodobenzene, 2-Bromoiodobenzene
Stabilization:
Stabilised with copper chip
B1126-25G
B1126-250G
B1126-5G
TCIAB1126-25GEA
353
DKK
TCIAB1126-25G
TCIAB1126-250G
TCIAB1126-5G
1-Brom-2-iodbenzen ≥98.0% (by GC) stabilised
1-Brom-2-iodbenzen
Formel:
C₆H₄BrI Molekylvægt: 282.91 g/mol Kogepunkt: 120°C |
MDL Number:
MFCD00001030 CAS-nummer: 583-55-1 EINECS: 209-508-9 |
|
Specifikationer Testresultater
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...