2-Methyl-2-propen-1-ol ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
3-Hydroxy-2-methylpropene, 2-Methyl-2-propen-1-ol, 2-Methyl allyl alcohol, 2-Methylprop-2-en-1-ol
M0091-25ML
M0091-5ML
M0091-500ML
TCIAM0091-25MLEA
386
DKK
TCIAM0091-25ML
TCIAM0091-5ML
TCIAM0091-500ML
2-Methyl-2-propen-1-ol ≥98.0% (by GC)
2-Methyl-2-propen-1-ol
Formel:
C₄H₈O Molekylvægt: 72.11 g/mol Kogepunkt: 113°C Flammepunkt: 33°C |
MDL Number:
MFCD00004737 CAS-nummer: 513-42-8 EINECS: 208-161-0 UN nummer: 2614 ADR: 3 III |
|
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...