3-Iodpropen ≥98.0% (by GC) stabilised

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig


Stabilization: Stabilised with copper chip

I0070-25G I0070-5G
TCIAI0070-25GEA 2180 DKK
TCIAI0070-25G TCIAI0070-5G
3-Iodpropen ≥98.0% (by GC) stabilised
3-Iodopropene
Formel: C₃H₅I
Kogepunkt: 102°C
Smeltepunkt: –99°C
Massefylde: 1,832 g/cm³ (20 °C)
Flammepunkt: 16°C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00001094
CAS-nummer: 556-56-9
EINECS: 209-130-4
UN nummer: 1723
ADR: 3,II
Merck Index: 14,00294
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=db514a546d8fff5ac4689b33f3d71d2f371caf60&vid=db514a546d8fff5ac4689b33f3d71d2f371caf60&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Brown to Dark purple clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR