3-Phenyl-1-propyn ≥95.0% (by GC) stabilised

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 2-Propynylbenzene

Stabilization: stabiliseret med BHT/Ionol

P1956-1G P1956-5G
TCIAP1956-1GEA 362 DKK
TCIAP1956-1G TCIAP1956-5G
3-Phenyl-1-propyn ≥95.0% (by GC) stabilised
3-Phenyl-1-propyn
Formel: C₉H₈
Kogepunkt: 75°C
Flammepunkt: 52°C
MDL Number: MFCD00134431
CAS-nummer: 10147-11-2
UN nummer: 3295
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7258d0c305b5234f07f80d99010523e42e0c3a68&vid=7258d0c305b5234f07f80d99010523e42e0c3a68&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR