Propagyl amin, HCl ≥98.0% (by total nitrogen and titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
P0990-25G
P0990-5G
TCIAP0990-25GEA
2230
DKK
TCIAP0990-25G
TCIAP0990-5G
Propagyl amin, HCl ≥98.0% (by total nitrogen and titration analysis)
Propagyl amin, HCl
Formel:
C₃H₆ClN Smeltepunkt: 178…179 °C |
MDL Number:
MFCD00012907 CAS-nummer: 15430-52-1 EINECS: 239-440-5 UN nummer: 2811 ADR: 6.1,III |
|
Specifikationer Testresultater
Appearance | White to Light yellow powder to crystal |
Purity (with Total Nitrogen) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Melting point | 175.0 to 180.0 deg-C |
Solubility in Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...