Rubrene ≥99.0% (by GC), purified by sublimation

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 5,6,11,12-Tetraphenylnaphthacene, 5,6,11,12-Tetraphenyltetracene

T2233-1G T2233-250MG
TCIAT2233-1GEA 1560 DKK
TCIAT2233-1G TCIAT2233-250MG
Rubrene ≥99.0% (by GC), purified by sublimation
Rubrene
Formel: C₄₂H₂₈
Smeltepunkt: 331°C
MDL Number: MFCD00003703
CAS-nummer: 517-51-1
EINECS: 208-242-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a327e93b62cb28dc86f35003e48c32a69762fa46&vid=eb4ac8be840a321473efd01ca18122ddf9ac092a&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR