Iodethan ≥99.0% (by GC) stabilised

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Ethyl iodide

Stabilization: Stabilised with copper chip

I0058-25G I0058-500G I0058-100G
TCIAI0058-25GEA 140 DKK
TCIAI0058-25G TCIAI0058-500G TCIAI0058-100G
Iodethan ≥99.0% (by GC) stabilised
Iodoethane
Formel: C₂H₅I
Kogepunkt: 72°C
Smeltepunkt: –108 °C
Massefylde: 1,936 g/cm³ (20 °C)
Flammepunkt: 53 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00001091
CAS-nummer: 75-03-6
EINECS: 200-833-1
UN nummer: 1993
ADR: 3,III
Merck Index: 14,03813
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7e48e4bcb15ac1ed1ccf70456be7db3a395e1a2a&vid=f37759a6600e2984fc741960a8d92b8af8b35ba0&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR