1-Iodbutan ≥98.0% (by GC) stabilised
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Stabilization:
Stabilised with copper chip
I0055-25G
I0055-500G
TCIAI0055-25GEA
238
DKK
TCIAI0055-25G
TCIAI0055-500G
1-Iodbutan ≥98.0% (by GC) stabilised
1-Iodobutane
Formel:
C₄H₉I Kogepunkt: 130°C Smeltepunkt: –103 °C Massefylde: 1,616 g/cm³ (20 °C) Flammepunkt: 49°C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00001098 CAS-nummer: 542-69-8 EINECS: 208-824-4 UN nummer: 1993 ADR: 3,III Merck Index: 14,01573 |
|
Specifikationer Testresultater
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...