1-Bromperfluoroctan ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

H0946-25G H0946-5G
TCIAH0946-25GEA 1470 DKK
TCIAH0946-25G TCIAH0946-5G
1-Bromperfluoroctan ≥98.0% (by GC)
1-Bromperfluoroctan
Formel: BrF₂C(CF₂)₆CF₃
Kogepunkt: 141°C
Smeltepunkt: 6 °C
Massefylde: 1,918 g/cm³ (25 °C)
MDL Number: MFCD00042082
CAS-nummer: 423-55-2
EINECS: 207-028-4
Merck Index: 14,07156
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b07ad74fdaaa9cb0bcc0b80352784500c515f866&vid=11933395b04d38ae4918fb8a57f33293fd84aaa0&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR