2-Octen cis- and trans-mixture ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

O0042-25ML O0042-500ML
TCIAO0042-25MLEA 378 DKK
TCIAO0042-25ML TCIAO0042-500ML
2-Octen cis- and trans-mixture ≥98.0% (by GC)
2-Octene
Formel: C₈H₁₆
Molekylvægt: 112.22 g/mol
Kogepunkt: 125°C
Smeltepunkt: -106.8
Flammepunkt: 21°C
MDL Number: MFCD00009532
CAS-nummer: 111-67-1
EINECS: 203-894-2
UN nummer: 3295
ADR: 3 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6d27e5737540d6642ac49357d7d30b6dcf1063dc&vid=cccf445718dd8781895589e3c1136bb3a8783208&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 % (total of isomers)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR