Calciumsulfat, Drierite™ 6 mesh for drying, with indicator (cobalt (II) chloride)
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Farlig
Synonymer: Calcium sulphate (anhydrous) anhydrite, Gypsum
43064.TD 43064.XA
43064.TDEA 676 DKK
43064.TD 43064.XA
Calciumsulfat, Drierite™ 6 mesh for drying, with indicator (cobalt (II) chloride)
Calcium sulphate
CAS No.: 7778-18-9 Physical Form: Granular, -6 Mesh Sensitivity: Hygroscopic Appearance: Blue to pink Odour: Odourless Merck Reference: 14,1706 MDL No.: MFCD00010912
Formel: CaO₄S
Molekylvægt: 136.14 g/mol
Smeltepunkt: 1350
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00010912
CAS-nummer: 7778-18-9
EINECS: 231-900-3
UN nummer: 3077
ADR: 9 III
Merck Index: 14,1706