4-Vinylphenylboronsyre (contains varying amounts of Anhydride)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Dihydroxy(4-vinylphenyl)borane, (4-Vinylphenyl)metaboric acid, 4-Vinylbenzeneboronic acid

V0075-1G V0075-5G
TCIAV0075-1GEA 214 DKK
TCIAV0075-1G TCIAV0075-5G
4-Vinylphenylboronsyre (contains varying amounts of Anhydride)
4-Vinylphenylboronsyre
Formel: C₈H₉BO₂
MDL Number: MFCD00239441
CAS-nummer: 2156-04-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd4faee154a5a6020cce4efb02ff644b45c3a9a4&vid=fd4faee154a5a6020cce4efb02ff644b45c3a9a4&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 114.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR