1,4-Phenyldiboronsäure (contains varying amounts of Anhydride)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: p-Phenylenebisboronic acid, 1,4-Phenylenediboronic acid, 1,4-Phenyldiboric acid, 1,4-Benzenediboronic acid

P1358-1G P1358-25G P1358-5G
TCIAP1358-1GEA 181 DKK
TCIAP1358-1G TCIAP1358-25G TCIAP1358-5G
1,4-Phenyldiboronsäure (contains varying amounts of Anhydride)
1,4-Phenyldiboronsäure
Formel: C₆H₈B₂O₄
MDL Number: MFCD00236018
CAS-nummer: 4612-26-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c139fa33642eecd1301d7ae3ad80bbf6e47739db&vid=c139fa33642eecd1301d7ae3ad80bbf6e47739db&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (Neutralization titration) 97.0 to 112.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR