Citral dimethyl acetal cis- and trans-mixture ≥90.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 1,1-Dimethoxy-3,7-dimethyl-2,6-octadiene

C1211-25ML C1211-500ML
TCIAC1211-25MLEA 255 DKK
TCIAC1211-25ML TCIAC1211-500ML
Citral dimethyl acetal cis- and trans-mixture ≥90.0% (by GC)
Citral dimethyl acetal
Formel: C₁₂H₂₂O₂
Molekylvægt: 198.31 g/mol
Kogepunkt: 103°C
Flammepunkt: 82°C
MDL Number: MFCD00008909
CAS-nummer: 7549-37-3
EINECS: 231-434-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=02447d2f151a244412717d7d0accfd74488ca485&vid=3f3ef2aefaff7f1cf65b7bfe98732865a03447ba&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) >90.0% (total of isomers)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR