5-Methylphenazinium methyl sulfat ≥98.0% (by HPLC, total nitrogen)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
PMS, 5-Methylphenazin-5-ium methyl sulphate, Phenazine methosulphate, Phenazine methylsulphate
P0083-1G
P0083-5G
TCIAP0083-1GEA
206
DKK
TCIAP0083-1G
TCIAP0083-5G
5-Methylphenazinium methyl sulfat ≥98.0% (by HPLC, total nitrogen)
5-Methylphenazinium methyl sulfat
Formel:
C₁₄H₁₄N₂O₄S Smeltepunkt: 160°C Opbevaringstemperatur: Fryser |
MDL Number:
MFCD00011923 CAS-nummer: 299-11-6 EINECS: 206-072-1 Merck Index: 14,06109 |
|
Specifikationer Testresultater
Appearance | Yellow to Brown to Dark green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...