(S)-(+)-1-Phenyl-1,2-ethandiol ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: (S)-(+)-Phenyl-1,2-ethanediol, (S)-1-Phenylethane-1,2-diol, (S)-(+)-Phenylethane-1,2-diol, (S)-(+)-α,β-Dihydroxyethylbenzene

P1151-1G P1151-5G
TCIAP1151-1GEA 132 DKK
TCIAP1151-1G TCIAP1151-5G
(S)-(+)-1-Phenyl-1,2-ethandiol ≥98.0% (by GC)
(S)-(+)-1-Phenyl-1,2-ethandiol
Formel: C₈H₁₀O₂
Kogepunkt: 82°C
Smeltepunkt: 67 °C
MDL Number: MFCD00066256
CAS-nummer: 25779-13-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ca693d973f51d872010b8dc310195fb425486e10&vid=ca693d973f51d872010b8dc310195fb425486e10&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 65.0 to 69.0 deg-C
Specific rotation [a]20/D +37.0 to +42.0 deg (C=3, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR