Siliciumstandard, 1g/l Si i vand AVS TITRINORM standard til AAS
Leverandør: VWR Chemicals
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
86710.180EA
255
DKK
86710.180
86710.260
Siliciumstandard, 1g/l Si i vand AVS TITRINORM standard til AAS
Standarder
Kationiske standarder
Silicium Standards
- Traceable to SRM from NIST
- Manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
- Supplied with certificate of analysis, real value ±0,3%