4-Nitro-2-picolin-1-oxid ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
2-Methyl-4-nitropyridine-1-oxide, 4-Nitro-2-picoline N-oxide, 2-Methyl-4-nitropyridine-N-oxide
M2114-25G
M2114-5G
TCIAM2114-25GEA
1350
DKK
TCIAM2114-25G
TCIAM2114-5G
4-Nitro-2-picolin-1-oxid ≥98.0% (by GC, titration analysis)
4-Nitro-2-picolin-1-oxid
Formel:
C₆H₆N₂O₃ Molekylvægt: 154.13 g/mol Smeltepunkt: 156 |
MDL Number:
MFCD00014625 CAS-nummer: 5470-66-6 |
|
Specifikationer Testresultater
Appearance | Light yellow to Yellow to Orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 154.0 to 158.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...