Yttrium 1.000 mg/l i fortyndet salpetersyre AVS TITRINORM standard til AAS
Leverandør: VWR Chemicals
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
86723.180EA
245
DKK
86723.180
86723.260
Yttrium 1.000 mg/l i fortyndet salpetersyre AVS TITRINORM standard til AAS
Standarder
Kationiske standarder
Yttrium Standards
- Traceable to SRM from NIST
- Manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
- Supplied with certificate of analysis, real value ±0,3%