Svovl 1.000 mg/l i vand AVS TITRINORM standard til AAS
Leverandør: VWR Chemicals
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
86706.180EA
219
DKK
86706.180
86706.260
Svovl 1.000 mg/l i vand AVS TITRINORM standard til AAS
Standarder
Kationiske standarder
Sulphur Standards
- Traceable to SRM from NIST
- Manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
- Supplied with certificate of analysis, real value ±0,3%