3-Nitro-5-(trifluormethyl)benzoesyre

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

T1344-25G T1344-5G
TCIAT1344-25GEA 3270 DKK
TCIAT1344-25G TCIAT1344-5G
3-Nitro-5-(trifluormethyl)benzoesyre
3-Nitro-5-(trifluormethyl)benzoesyre
Formel: C₈H₄F₃NO₄
MDL Number: MFCD00024509
CAS-nummer: 328-80-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2432091700eca3df39dab96246a6396bf80b315f&vid=2432091700eca3df39dab96246a6396bf80b315f&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Purity (Neutralization titration) min. 98.0 %
Melting point 128.0 to 132.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR