Chlormethyl pivalat ≥99.0% (by GC) amino-protecting agent
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
Chloromethyl trimethylacetate, Trimethylacetic acid chloromethyl ester, 2,2-Dimethylpropionic acid chloromethyl ester, Pivalic acid chloromethyl ester
P1012-25G
P1012-500G
P1012-100G
TCIAP1012-25GEA
156
DKK
TCIAP1012-25G
TCIAP1012-500G
TCIAP1012-100G
Chlormethyl pivalat ≥99.0% (by GC) amino-protecting agent
Chlormethyl pivalat
Formel:
C₆H₁₁ClO₂ Kogepunkt: 147°C Massefylde: 1,0387 g/cm³ (20 °C) Flammepunkt: 40°C |
MDL Number:
MFCD00000884 CAS-nummer: 18997-19-8 EINECS: 242-735-1 UN nummer: 2924 ADR: 3,III |
|
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...