3-Mercapto-1,2-propandiol 95,0 GC_ASSAY_METHOD
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
alpha-Thioglycerol, α-Monothioglycerol, Monothioglycerol, (±)-3-Mercapto-1,2-dihydroxy propane, 1-Thioglycerol, (±)-α-Monothioglycerine, (±)-1-Thioglycerine, (±)-3-Mercapto-1,2-propanediol
T0905-25G
T0905-500G
T0905-100G
TCIAT0905-25GEA
148
DKK
TCIAT0905-25G
TCIAT0905-500G
TCIAT0905-100G
3-Mercapto-1,2-propandiol 95,0 GC_ASSAY_METHOD
3-Mercapto-1,2-propandiol
Formel:
C₃H₈O₂S Molekylvægt: 108.16 g/mol Kogepunkt: 139°C Flammepunkt: 113°C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00004879 CAS-nummer: 96-27-5 EINECS: 202-495-0 UN nummer: 2810 ADR: 6.1 III Merck Index: 14,09335 |
|
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...