4-Bromphenylboronsyre (contains varying amounts of Anhydride)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 4-Bromobenzeneboronic acid, (4-Bromophenyl)boronic acid, Dihydroxy(4-bromophenyl)borane, (4-Bromophenyl)metaboric acid

B1858-1G B1858-25G B1858-5G
TCIAB1858-1GEA 132 DKK
TCIAB1858-1G TCIAB1858-25G TCIAB1858-5G
4-Bromphenylboronsyre (contains varying amounts of Anhydride)
4-Bromphenylboronsyre
Formel: C₆H₆BBrO₂
Massefylde: 1,67 g/cm³ (20 °C)
MDL Number: MFCD00002104
CAS-nummer: 5467-74-3
EINECS: 226-779-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d037b74423b4a23efbaa90db9c84801eeaec87c7&vid=d037b74423b4a23efbaa90db9c84801eeaec87c7&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (Neutralization titration) 97.0 to 110.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR