2-Fluorphenylboronsyre (contains varying amounts of Anhydride)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Dihydroxy(2-fluorophenyl)borane, 2-Fluorobenzeneboronic acid, (2-Fluorophenyl)metaboric acid, (2-Fluorophenyl)boronic acid

F0407-1G F0407-25G F0407-5G
TCIAF0407-1GEA 263 DKK
TCIAF0407-1G TCIAF0407-25G TCIAF0407-5G
2-Fluorphenylboronsyre (contains varying amounts of Anhydride)
2-Fluorphenylboronsyre
Formel: C₆H₆BFO₂
MDL Number: MFCD00674013
CAS-nummer: 1993-03-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cf2fcd11d3a13907d9848385f75d7ff488e698ef&vid=cf2fcd11d3a13907d9848385f75d7ff488e698ef&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (Neutralization titration) 97.0 to 115.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR