Homovanillinsyre ≥98.0% (by titrimetric analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
2-(4-Hydroxy-3-methoxyphenyl)acetic acid, (4-Hydroxy-3-methoxyphenyl)acetic acid
H0339-1G
H0339-25G
H0339-5G
TCIAH0339-1GEA
255
DKK
TCIAH0339-1G
TCIAH0339-25G
TCIAH0339-5G
Homovanillinsyre ≥98.0% (by titrimetric analysis)
Homovanillinsyre
Formel:
(C₆H₃(OH)(OCH₃))CH₂COOH Kogepunkt: 339 °C (1013 hPa) Smeltepunkt: 145 °C |
MDL Number:
MFCD00004350 CAS-nummer: 306-08-1 EINECS: 206-176-7 Merck Index: 14,04740 |
Specifikationer Testresultater
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 141.0 to 144.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...