3-(Brommethyl)pentan ≥97.0% (by GC) stabilised

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 3-(Bromomethyl)pentane

Stabilization: Stabilised with copper chip

B1905-25G B1905-5G
TCIAB1905-25GEA 813 DKK
TCIAB1905-25G TCIAB1905-5G
3-(Brommethyl)pentan ≥97.0% (by GC) stabilised
1-Bromo-2-ethylbutane
Formel: C₆H₁₃Br
Kogepunkt: 145°C
Massefylde: 1,179 g/cm³ (30 °C)
Flammepunkt: 36°C
MDL Number: MFCD00000219
CAS-nummer: 3814-34-4
EINECS: 223-302-6
UN nummer: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a7ea9a3c9b3bd08d7fdc1019be3e84d0a83136aa&vid=a7ea9a3c9b3bd08d7fdc1019be3e84d0a83136aa&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR