3a,4,7,7a-Tetrahydro-4,7-methanoinden ≥97.0% (by GC) stabilised precursor to cyclopentadiene
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Stabilization:
stabiliseret med BHT/Ionol
D0443-25ML
D0443-500ML
TCIAD0443-25MLEA
132
DKK
TCIAD0443-25ML
TCIAD0443-500ML
3a,4,7,7a-Tetrahydro-4,7-methanoinden ≥97.0% (by GC) stabilised precursor to cyclopentadiene
3a,4,7,7a-Tetrahydro-4,7-methanoinden
Formel:
C₁₀H₁₂ Kogepunkt: 170 °C (1013 hPa) Smeltepunkt: 12…34 °C Massefylde: 0,982 g/cm³ (20 °C) Flammepunkt: 44°C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00078246 CAS-nummer: 77-73-6 EINECS: 201-052-9 UN nummer: 2048 ADR: 3,III Merck Index: 13,02767 |
|
Specifikationer Testresultater
Appearance | White or Colorless to Almost white or Almost colorless powder to lump to clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...