(2-Cyanoethyl)triethoxysilan ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Triethoxy(2-cyanoethyl)silane, 3-(Triethoxysilyl)propiononitrile, 3-(Triethoxysilyl)propionitrile
C1259-25ML
C1259-250ML
TCIAC1259-25MLEA
222
DKK
TCIAC1259-25ML
TCIAC1259-250ML
(2-Cyanoethyl)triethoxysilan ≥98.0% (by GC)
(2-Cyanoethyl)triethoxysilan
Formel:
NCCH₂CH₂Si(OCH₂CH₃)₃ Molekylvægt: 217.34 g/mol Kogepunkt: 137°C Flammepunkt: 86°C |
MDL Number:
MFCD00019851 CAS-nummer: 919-31-3 EINECS: 213-050-5 |
|
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...