2-Chlorcyclopentanon ≥95.0% (by GC) stabilised
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Stabilization:
Stabilised with hydroquinone (HQ) and calcium carbonate
C1437-25G
C1437-5G
TCIAC1437-25GEA
2630
DKK
TCIAC1437-25G
TCIAC1437-5G
2-Chlorcyclopentanon ≥95.0% (by GC) stabilised
2-Chlorocyclopentanone
Formel:
C₅H₇ClO Kogepunkt: 90°C Massefylde: 1,2061 g/cm³ (0 °C) Flammepunkt: 77°C Opbevaringstemperatur: Fryser |
MDL Number:
MFCD00001410 CAS-nummer: 694-28-0 EINECS: 211-769-9 ADR: 6.1,III |
|
Specifikationer Testresultater
Appearance | White to Yellow to Green clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...