Bronopol ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 2-Brom-2-nitropropan-1,3-diol

B1247-25G B1247-500G B1247-100G
TCIAB1247-25GEA 189 DKK
TCIAB1247-25G TCIAB1247-500G TCIAB1247-100G
Bronopol ≥98.0% (by titrimetric analysis)
Bronopol
Formel: HOCH₂BrC(NO₂)CH₂OH
Molekylvægt: 199.99 g/mol
Kogepunkt: 388
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00007390
CAS-nummer: 52-51-7
EINECS: 200-143-0
UN nummer: 3241
ADR: 4.1 III
Merck Index: 14,01447
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8974c292739dfbaa734477c6902f128f8fbc41fb&vid=8974c292739dfbaa734477c6902f128f8fbc41fb&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (Argentmetric Titration) min. 98.0 %
Melting point 130.0 to 133.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR