Capillary columns, OPTIMA® 1701
Leverandør: MACHEREY-NAGEL
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
726824.25 726824.30 726824.50 726824.60 726841.25 726841.50 726929.25 726929.30 726929.50 726929.60 726965.30 726058.10 726058.15 726058.25 726058.30 726058.50 726058.60 726064.30 726064.60 726318.10 726318.15 726318.25 726318.30 726318.50 726318.60 726320.25 726320.30 726320.50 726320.60 726545.10 726545.15 726545.25 726545.30 726735.15 726735.25 726735.30 726735.50
MANA726824.25EA 9030 DKK
MANA726824.25 MANA726824.30 MANA726824.50 MANA726824.60 MANA726841.25 MANA726841.50 MANA726929.25 MANA726929.30 MANA726929.50 MANA726929.60 MANA726965.30 MANA726058.10 MANA726058.15 MANA726058.25 MANA726058.30 MANA726058.50 MANA726058.60 MANA726064.30 MANA726064.60 MANA726318.10 MANA726318.15 MANA726318.25 MANA726318.30 MANA726318.50 MANA726318.60 MANA726320.25 MANA726320.30 MANA726320.50 MANA726320.60 MANA726545.10 MANA726545.15 MANA726545.25 MANA726545.30 MANA726735.15 MANA726735.25 MANA726735.30 MANA726735.50
Capillary columns, OPTIMA® 1701
Kromatografikolonner
Medium polar GC phase.
  • Special selectivity due to high cyanopropyl content
  • Reference column for structure identification in combination with column of polarity 5 (e.g., OPTIMA® 5)
  • Maximum temperature for isothermal operation 280 °C
  • Maximum temperature for short isotherms in a temperature program 300 °C
14% cyanopropyl-phenyl – 86% dimethylpolysiloxane phase.
USP code: G46.