Test tubes, with rim, medium wall
Leverandør: MURRAY S
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
G022/31 G022/33 G022/34 G022/35 G022/38 G022/40 G022/32
212-0560EA 412 DKK
212-0560 212-0456 212-0561 212-0562 212-0563 212-0564 212-0455
Test tubes, with rim, medium wall
Rør Reagensglas
Neutral glass, SAMCO '50'
  • Medium walled
  • Withstand moderate temperature change
Certificeringer: Manufactured to the dimensions, tolerances and quality requirements of ISO 4142:2002, to meet all applications for re-usable glass tubes.
Advarsel: Tubes should not be taken from cold and placed directly into the hottest part of the flame without pre-heating.