Millex® Syringe Filters, PVDF Membrane, Sterile
Leverandør: Merck
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
SLSV025LS SLGV033RS SLGV013SL SLHV013SL SLHV033RS SLHV033RB SLVV033RS SLGV033RB SLGVM33RS SLHVM33RS SLGVV255F SLGV0250S
514-0452EA 2000 DKK
514-0452 MILFSLGV033RS 514-0427 514-0439 MILFSLHV033RS MILFSLHV033RB MILFSLVV033RS MILFSLGV033RB MILFSLGVM33RS MILFSLHVM33RS MILFSLGVV255F MILFSLGV0250S
Millex® Syringe Filters, PVDF Membrane, Sterile
Filtre Sprøjtefiltre
Filters for sterilising or clarifying biological solutions. The larger filtration area increases flow rate and throughput. It also makes it easier to filter solutions because it reduces the pressure required to empty the syringe.
Manufacturing occurs in a controlled environment using an automated process. Provided with a certificate of quality.