Chemical resistant gloves, AlphaTec® 87-600
Leverandør: Ansell
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
87-600/8 87-600/9 87-600/7 87-600/9.5-10
ANSE87-600/8EA 0 DKK
ANSE87-600/8 ANSE87-600/9 ANSE87-600/7 ANSE87-600/9.5-10
Chemical resistant gloves, AlphaTec® 87-600
Handsker
These unflocked natural latex gloves provide strength and elasticity for the safe handling of more hazardous substances.
  • Only 0,43 mm thick for outstanding sensitivity and tactile performance
  • Reduced risk of allergic reactions
  • Fishscale finish for improved grip
Ideal for use in pharmaceutical industries, precision assembly, cleaning, maintenance and for janitorial use. Also suitable for handling fibreglass.
Certificeringer: EN 421, EN 388:2016 (X010X), EN ISO 374-1:2016 Type B (KLT), EN ISO 374-5:2016; AQL 0,65; Cat. III