Safety boots, Wellingtons, Steelite™ Food FW 84
Leverandør: Portwest
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
FW84WHR42 FW84WHR41 FW84WHR47 FW84WHR46 FW84WHR45 FW84WHR44 FW84WHR43 FW84WHR40 FW84WHR39 FW84WHR38 FW84WHR37 FW84WHR36 FW84WHR48
114-3660EA 301 DKK
114-3660 114-3659 114-3665 114-3664 114-3663 114-3662 114-3661 114-3658 114-3657 114-3656 114-3655 114-7837 PRTWFW84WHR48
Safety boots, Wellingtons, Steelite™ Food FW 84
Sko og støvler
These anti-static, waterproof safety wellingtons are manufactured from a PVC/nitrile upper with a PVC/nitrile sole. Boots are resistant to oils, fats, alkalis, and acids.
  • Washable nylon lining
  • Shock-absorbent heel
  • Protective steel toecap (200 J)
  • Kick-off spurs
Suitable for food industry.
Colour: White.
Certificeringer: EN ISO 20345:2011; S4, SRC.