Anion exchange chromatography columns, TSK-GEL®, sugar
Leverandør: TOSOH Bioscience
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
0008639 0008640
TOYO0008639EA 13340 DKK
TOYO0008639 TOYO0008640
Anion exchange chromatography columns, TSK-GEL®, sugar
Kromatografikolonner
TSK-GEL® sugar columns are polymer based stainless steel columns for analysis of monosacchrides, disaccharides and sugar alcohols by isocratic analysis (TSKgel sugar analysis AXI) or by gradient analysis (TSKgel sugar analysis AXG).