Erlenmeyer flasks, narrow or wide neck, beaded rim
Leverandør: Kavalier
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
632411119025 632417119100 632417119250 632417119500 632417106250 632417106500 632411119956 632411106025 632417106100 632417119940 632417119956 632417106200 632417119200 632417106940 632417106950 632417119300 632417119950
214-1300EA 2120 DKK
214-1300 214-1302 214-1304 214-1306 214-1314 214-1316 214-1322 214-1331 214-1333 KAVA632417119940 KAVA632417119956 KAVA632417106200 KAVA632417119200 KAVA632417106940 KAVA632417106950 KAVA632417119300 KAVA632417119950
Erlenmeyer flasks, narrow or wide neck, beaded rim
Kolber Erlenmeyer-kolber
Made of borosilicate glass 3.3., clear.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Volume graduations and labelling area.