Ribbons (heavy-duty), for BBP™31 printers
Leverandør: Brady
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
118085 118089 118084 118090 118094 118093 118091 118086 118088 118097 118923 133357
BRDY118085EA 1590 DKK
BRDY118085 BRDY118089 BRDY118084 BRDY118090 BRDY118094 116-0416 BRDY118091 BRDY118086 BRDY118088 BRDY118097 BRDY118923 BRDY133357
Ribbons (heavy-duty), for BBP™31 printers
Printer bånd
'Drop and Click' print ribbons for quick and easy supply changes.
  • Tough, smear-proof ribbons for long-lasting printed text
  • Print can withstand years in industrial areas and outdoor conditions
  • No calibrating, no threading, no waste, no hassle
Width: 110 mm