Trenbolone, Supelco®

Leverandør: Merck
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

T3925-250MG T3925-1G
T3925-250MGEA 1340 DKK
T3925-250MG T3925-1G
Trenbolone, Supelco®
Trenbolone
Formel: C₁₈H₂₂O₂
MDL Number: MFCD00214395
CAS-nummer: 10161-33-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d76fddbeca1fe143524f4a1858afafdecfa322e0&vid=a7b906d0f2624cdcdd3fb31c670964ff535e13a7&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=965a3e1bb952689e40931c3466a3f643217e6aea&vid=53c2392d2eb1e09ec6d45d99e608dde06f9c8638&prid=web&clid=SAPDAM

Bestil nu

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR