(R)-(+)-α-Lipoic acid ≥98.0% (by HPLC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
(R)-5-(1,2-Dithiolan-3-yl)pentanoic acid, (R)-alpha-Lipoic acid, (R)-1,2-Dithiolane-3-valeric acid, (R)-Thioctic acid
L0207-25G
L0207-5G
L0207-100G
TCIAL0207-25GEA
1680
DKK
TCIAL0207-25G
TCIAL0207-5G
TCIAL0207-100G
(R)-(+)-α-Lipoic acid ≥98.0% (by HPLC, titration analysis)
(R)-(+)-α-Lipoic acid
Formel:
C₈H₁₄O₂S₂ Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD01631142 CAS-nummer: 1200-22-2 Merck Index: 14,09326 |
|
Specifikationer Testresultater
Appearance | Light orange to Yellow to Green powder to crystaline |
Purity (HPLC) (CAD) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Specific rotation [a]20/D | +110 to +121 deg (C=1, EtOH) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...