(±)-α-Lipoic acid ≥99.0% (by titrimetric analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
5-(1,2-Dithiolan-3-yl)pentanoic acid, (±)-1,2-Dithiolane-3-pentanoic acid, 5-(1,2)Dithiolan-3-yl-pentanoic acid, DL-α-Lipoic acid, DL-Thioctic acid (oxidized), DL-6,8-Thioctic acid, Lip(S2), 6,8-Dithiooctanoic acid
L0058-25G
L0058-5G
TCIAL0058-25GEA
1290
DKK
TCIAL0058-25G
TCIAL0058-5G
(±)-α-Lipoic acid ≥99.0% (by titrimetric analysis)
(±)-α-Lipoic acid
Formel:
C₈H₁₄O₂S₂ Kogepunkt: 165°C Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD00005474 CAS-nummer: 1077-28-7 EINECS: 214-071-2 Merck Index: 14,09326 |
|
Specifikationer Testresultater
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...